Assay Details
| Gene Symbol: Clk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027853.1 | 3 - 4 | 561 | 111 | ||
| GenBank mRNA | BC145905.1 | - | 3 - 4 | 464 | 111 | |
| BC145911.1 | - | 3 - 4 | 464 | 111 | ||
| L29221.1 | - | 3 - 4 | 477 | 111 | ||
| U21209.1 | - | 3 - 4 | 477 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Clk1
Genomic Map