Assay Details
| Gene Symbol: Cxcr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB003174.1 | - | 1 - 2 | 103 | 58 | |
| BC096626.1 | - | 1 - 2 | 76 | 30749408 | 58 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB003174.1,BC096626.1 transcripts.
Set Membership: |
|

Cxcr3
Genomic Map