Assay Details
| Gene Symbol: Camp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009921.2 | NP_034051.2 | 1 - 2 | 219 | 69 | |
| GenBank mRNA | AK158005.1 | - | 1 - 2 | 211 | 69 | |
| BC120752.1 | - | 1 - 2 | 216 | 40129675 | 69 | |
| BC120754.1 | - | 1 - 2 | 219 | 40129677 | 69 | |
| U43409.1 | - | 1 - 2 | 212 | 69 | ||
| X94353.1 | - | 1 - 2 | 211 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Camp
Genomic Map