Assay Details
| Gene Symbol: Cox4i1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293559.1 | NP_001280488.1 | 4 - 5 | 428 | 74 | |
| NM_009941.3 | NP_034071.2 | 4 - 5 | 503 | 74 | ||
| GenBank mRNA | AK002943.1 | - | 760 | 74 | ||
| AK012583.1 | - | 4 - 5 | 440 | 74 | ||
| AK019276.1 | - | 4 - 5 | 405 | 74 | ||
| AK150447.1 | - | 4 - 5 | 433 | 74 | ||
| BC132269.1 | - | 4 - 5 | 424 | 40130546 | 74 | |
| BC132275.1 | - | 4 - 5 | 424 | 40130552 | 74 | |
| M37829.1 | - | 4 - 5 | 426 | 74 | ||
| X54691.1 | - | 4 - 5 | 428 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26221739 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26221738 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26221741 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26221737 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P165435 | M37829.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P165435 | X54691.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P165435 | AK002943.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P165435 | AK012583.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P165435 | NM_009941.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P165435 | M37829.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P165435 | X54691.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P165435 | AK002943.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P165435 | AK012583.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P165435 | NM_009941.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 288681 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 43175 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 394334 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 93299 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P125985 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P165435 | M37829.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P165435 | X54691.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P165435 | AK002943.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P165435 | AK012583.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P165435 | NM_009941.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P165435 | M37829.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P165435 | X54691.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P165435 | AK002943.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P165435 | AK012583.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P165435 | NM_009941.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P165435 | M37829.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P165435 | X54691.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P165435 | AK002943.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P165435 | AK012583.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P165435 | NM_009941.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P165435 | M37829.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P165435 | X54691.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P165435 | AK002943.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P165435 | AK012583.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P165435 | NM_009941.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26221752 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26221744 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26221766 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26221772 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26221762 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26221763 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26221739 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26221745 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26221738 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26221758 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26221764 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26221749 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26221750 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26221747 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26221765 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26221753 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26221743 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26221759 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26221742 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26221774 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26221777 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26221756 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26221741 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26221775 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26221768 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P125985 | - | - |
| Whole Mouse Genome G4122A | A_51_P165435 | M37829.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P165435 | X54691.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P165435 | AK002943.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P165435 | AK012583.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P165435 | NM_009941.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P165435 | M37829.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P165435 | X54691.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P165435 | AK002943.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P165435 | AK012583.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P165435 | NM_009941.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111194 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P125985 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07359886 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26221738 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26221739 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26221737 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26221741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26221778 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26221830 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26221749 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06848919 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26221742 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26221752 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07215706 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07368896 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26221787 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07368908 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26221745 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26221825 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26221741 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26221759 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07368902 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26221766 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26221822 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26221753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07368905 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26221768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26221821 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26221829 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07215708 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26221831 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26221783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26221823 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26221777 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26221832 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26221774 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26221764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26221744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26221743 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26221835 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26221786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06854807 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26221778 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26221775 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26221750 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10576032 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10576033 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10576030 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10576031 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448322_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10576029 | - | - |
| Mouse Genome 430 2.0 Array | 1448322_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1448322_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1434952_at | - | - |
| Mouse Genome 430 2.0 Array | 1434952_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12857_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12857_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800011844.mm.1 | NM_009941.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011844.mm.1 | TC0800001447.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800011846.mm.1 | NM_009941.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011846.mm.1 | TC0800001447.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800011842.mm.1 | NM_009941.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011842.mm.1 | TC0800001447.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800011845.mm.1 | NM_009941.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011845.mm.1 | TC0800001447.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448322_PM_a_at | - | - |
| Mouse Expression Set 430 | 1448322_a_at | - | - |
| Murine Genome U74v2 Set | 102124_f_at | - | - |
| Murine Genome U74v2 Set | 93788_PM_at | - | - |
| Murine Genome U74v2 Set | 102124_PM_f_at | - | - |
| Murine Genome U74v2 Set | 93788_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448322_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448322_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448322_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448322_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 910978 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5340427 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005340427 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1249867 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1249867 | - | - |
| mouse-6 v1.1 (Illumina) | scl33243.5.1_27-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5340427 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl33243.5.1_27-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5340427 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2600274 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2600274 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl33243.5.1_27-S | - | - |

Cox4i1
Genomic Map