Assay Details
| Gene Symbol: Cradd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009950.2 | NP_034080.1 | 1 - 2 | 117 | 107 | |
| GenBank mRNA | AJ224738.1 | - | 1 - 2 | 116 | 106 | |
| AK163112.1 | - | 1 - 2 | 129 | 107 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AJ224738.1,AK163112.1,NM_009950.2 transcripts.
Set Membership: |
|

Cradd
Genomic Map