Assay Details
| Gene Symbol: Cst7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009977.3 | NP_034107.2 | 1 - 2 | 173 | 81 | |
| GenBank mRNA | AB015224.1 | - | 1 - 2 | 154 | 81 | |
| AF031825.1 | - | 1 - 2 | 102 | 81 | ||
| AK004420.1 | - | 1 - 2 | 174 | 81 | ||
| AK170583.1 | - | 1 - 2 | 120 | 81 | ||
| BC117047.1 | - | 1 - 2 | 158 | 40126366 | 81 | |
| BC117049.1 | - | 1 - 2 | 158 | 40126368 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cst7
Genomic Map