Assay Details
| Gene Symbol: Cx3cr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009987.4 | NP_034117.3 | 1 - 2 | 92 | 70 | |
| GenBank mRNA | AF102269.1 | - | 1 - 2 | 86 | 70 | |
| AK035902.1 | - | 1 - 2 | 67 | 70 | ||
| AK044557.1 | - | 1 - 2 | 93 | 70 | ||
| AK045634.1 | - | 1 - 2 | 80 | 70 | ||
| AK137388.1 | - | 1 - 2 | 80 | 70 | ||
| AK140010.1 | - | 1 - 2 | 66 | 70 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF102269.1,AK035902.1,AK044557.1,AK045634.1,AK137388.1,AK140010.1,NM_009987.4 transcripts.
Set Membership: |
|

Cx3cr1
Genomic Map