Assay Details
| Gene Symbol: Dnase2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010062.3 | NP_034192.1 | 2 - 3 | 428 | 62 | |
| GenBank mRNA | AK145192.1 | - | 2 - 3 | 429 | 62 | |
| AK167277.1 | - | 2 - 3 | 394 | 62 | ||
| BC058609.1 | - | 2 - 3 | 375 | 5397016 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dnase2a
Genomic Map