Assay Details
| Gene Symbol: Dvl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK155349.1 | - | 13 - 14 | 1811 | 70 | |
| AK162430.1 | - | 7 - 8 | 910 | 70 | ||
| AK220168.1 | - | 11 - 12 | 2084 | 70 | ||
| BC018593.1 | - | 6 - 7 | 695 | 70 | ||
| BC051991.1 | - | 11 - 12 | 1726 | 70 | ||
| BC088987.1 | - | 12 - 13 | 1746 | 70 | ||
| BC138848.1 | - | 14 - 15 | 1682 | 8861870 | 70 | |
| BC138849.1 | - | 14 - 15 | 1682 | 8861871 | 70 | |
| U10115.1 | - | 13 - 14 | 1768 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dvl1
Genomic Map