Assay Details
| Gene Symbol: Epas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010137.3 | NP_034267.3 | 10 - 11 | 1863 | 103 | |
| GenBank mRNA | AF045160.1 | - | 10 - 11 | 1627 | 103 | |
| AK136911.1 | - | 10 - 11 | 1867 | 103 | ||
| AK141185.1 | - | 10 - 11 | 1598 | 103 | ||
| AK158430.1 | - | 10 - 11 | 2027 | 103 | ||
| BC057870.1 | - | 10 - 11 | 1585 | 5032291 | 103 | |
| D89787.1 | - | 10 - 11 | 1825 | 103 | ||
| U81983.1 | - | 10 - 11 | 1546 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epas1
Genomic Map