Assay Details
| Gene Symbol: Ephb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159571.1 | NP_001153043.1 | 7 - 8 | 1938 | 73 | |
| NM_010144.6 | NP_034274.4 | 7 - 8 | 1911 | 73 | ||
| GenBank mRNA | AK046884.1 | - | 7 - 8 | 1876 | 73 | |
| AK049340.1 | - | 7 - 8 | 1912 | 73 | ||
| AK132392.1 | - | 7 - 8 | 1911 | 73 | ||
| AK143422.1 | - | 1881 | 73 | |||
| BC013342.1 | - | 7 - 8 | 1599 | 73 | ||
| BC016505.1 | - | 7 - 8 | 1863 | 3709926 | 73 | |
| BC090839.1 | - | 7 - 8 | 1688 | 73 | ||
| U06834.1 | - | 8 - 9 | 1524 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ephb4
Genomic Map