| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100016259.mm.1 |
NM_001252192.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100016259.mm.1 |
NM_001252192.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100016259.mm.1 |
NM_010164.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100016259.mm.1 |
NM_010164.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100016259.mm.1 |
TC0100001985.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100016257.mm.1 |
NM_010164.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100016257.mm.1 |
NM_010164.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100016257.mm.1 |
TC0100001985.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100016261.mm.1 |
NM_001252192.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100016261.mm.1 |
NM_001252192.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100016261.mm.1 |
NM_010164.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100016261.mm.1 |
NM_010164.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100016261.mm.1 |
TC0100001985.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100016262.mm.1 |
NM_001252192.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100016262.mm.1 |
NM_001252192.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100016262.mm.1 |
NM_010164.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100016262.mm.1 |
NM_010164.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100016262.mm.1 |
TC0100001985.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100016263.mm.1 |
NM_001252192.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100016263.mm.1 |
NM_001252192.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100016263.mm.1 |
NM_010164.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100016263.mm.1 |
NM_010164.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100016263.mm.1 |
TC0100001985.mm.1 |
MTA TC |