Assay Details
| Gene Symbol: Eya1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252192.1 | NP_001239121.1 | 5 - 6 | 894 | 71 | |
| GenBank mRNA | AK042412.1 | - | 5 - 6 | 817 | 71 | |
| U61110.1 | - | 5 - 6 | 567 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eya1
Genomic Map