Assay Details
| Gene Symbol: Eya4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010167.4 | NP_034297.2 | 2 - 3 | 241 | 69 | |
| GenBank mRNA | AK041625.1 | - | 2 - 3 | 242 | 69 | |
| BC120898.1 | - | 2 - 3 | 225 | 40061597 | 69 | |
| BC120899.1 | - | 2 - 3 | 225 | 40061599 | 69 | |
| Y17115.1 | - | 3 - 4 | 352 | 69 | ||
More Information
Set Membership: |
|

Eya4
Genomic Map