Assay Details
| Gene Symbol: Eya4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010167.4 | NP_034297.2 | 6 - 7 | 577 | 64 | |
| GenBank mRNA | AK041625.1 | - | 6 - 7 | 578 | 64 | |
| BC120898.1 | - | 6 - 7 | 561 | 40061597 | 64 | |
| BC120899.1 | - | 6 - 7 | 561 | 40061599 | 64 | |
| Y17115.1 | - | 7 - 8 | 688 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eya4
Genomic Map