Assay Details
| Gene Symbol: F2rl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010170.4 | NP_034300.3 | 1 - 2 | 210 | 72 | |
| GenBank mRNA | AK089444.1 | - | 1 - 2 | 174 | 72 | |
| AK135212.1 | - | 1 - 2 | 277 | 72 | ||
| BC140985.1 | - | 1 - 2 | 165 | 9055597 | 72 | |
| U92972.1 | - | 1 - 2 | 218 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P261164 | U92972.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P261164 | U92972.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P162507 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P261164 | U92972.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P261164 | U92972.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P261164 | U92972.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P261164 | U92972.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29242082 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29242061 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29242098 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29242069 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29242079 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29242094 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29242088 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29242075 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29242083 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29242104 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29242063 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29242070 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29242092 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29242059 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29242084 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29242058 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29242099 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29242095 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29242071 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29242086 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29242093 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29242067 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29242090 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29242065 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29242066 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29242102 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07960849 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P162507 | - | - |
| Whole Mouse Genome G4122A | A_51_P261164 | U92972.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P261164 | U92972.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129982 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P162507 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10500520 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10500514 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10500519 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10500516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29242119 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242056 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242115 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10849901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242119 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242059 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242108 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242123 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242050 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10958019 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242039 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242061 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242071 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242098 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242130 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242131 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242069 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242128 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242053 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242113 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10518991 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10500514 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242041 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242120 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10550419 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242106 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10500520 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242067 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242104 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242090 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242076 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242040 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242110 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11011902 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242095 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242044 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242083 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242088 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242037 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242066 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242079 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242036 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242054 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10518992 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242092 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10849902 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242102 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242077 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242126 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11011894 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242093 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242111 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10500519 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242057 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242091 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29242046 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10406739 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10406738 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10406737 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421407_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1445680_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10406736 | - | - |
| Mouse Genome 430 2.0 Array | 1421407_at | - | - |
| Mouse Genome 430 2.0 Array | 1445680_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1445680_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1421407_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14064_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14064_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300006879.mm.1 | NM_010170.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300006879.mm.1 | NM_010170.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300006879.mm.1 | TC1300001081.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300006877.mm.1 | NM_010170.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300006877.mm.1 | NM_010170.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300006877.mm.1 | TC1300001081.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300006878.mm.1 | NM_010170.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300006878.mm.1 | NM_010170.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300006878.mm.1 | TC1300001081.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300006881.mm.1 | NM_010170.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300006881.mm.1 | NM_010170.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300006881.mm.1 | TC1300001081.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300006876.mm.1 | NM_010170.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300006876.mm.1 | NM_010170.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300006876.mm.1 | TC1300001081.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300006880.mm.1 | NM_010170.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300006880.mm.1 | NM_010170.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300006880.mm.1 | TC1300001081.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300006875.mm.1 | NM_010170.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300006875.mm.1 | NM_010170.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300006875.mm.1 | TC1300001081.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1421407_at | - | - |
| Mouse Expression Set 430 | 1421407_PM_at | - | - |
| Mouse Expression Set 430 | 1445680_PM_x_at | - | - |
| Mouse Expression Set 430 | 1445680_x_at | - | - |
| Murine Genome U74v2 Set | 94762_at | - | - |
| Murine Genome U74v2 Set | 94762_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421407_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421407_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421407_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421407_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1445680_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1445680_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 380612 | - | - |
| ABI Mouse Genome Survey Microarray | 531037 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6110736 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6110736 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006110736 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1236465 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1236465 | - | - |
| mouse-6 v1.1 (Illumina) | scl44500.2_38-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6110736 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl44500.2_38-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6110736 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1236465 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1236465 | - | - |

F2rl2
Genomic Map