Assay Details
| Gene Symbol: Fcer1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010184.2 | NP_034314.2 | 3 - 4 | 374 | 148 | |
| GenBank mRNA | BC125455.1 | - | 3 - 4 | 339 | 40129970 | 148 |
| BC125457.1 | - | 3 - 4 | 338 | 40129972 | 148 | |
| BC145138.1 | - | 3 - 4 | 296 | 148 | ||
| CJ142154.1 | - | 3 - 4 | 295 | 148 | ||
| CJ142191.1 | - | 3 - 4 | 374 | 148 | ||
| J05018.1 | - | 3 - 4 | 359 | 148 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
positive regulation of type I hypersensitivity
serotonin secretion
signal transduction
cell surface receptor signaling pathway
activation of JUN kinase activity
leukotriene biosynthetic process
Fc-epsilon receptor signaling pathway
positive regulation of mast cell degranulation
positive regulation of interleukin-3 biosynthetic process
positive regulation of granulocyte macrophage colony-stimulating factor biosynthetic process
positive regulation of peptidyl-tyrosine phosphorylation
positive regulation of calcium-mediated signaling
protein binding
IgE receptor activity
IgE binding
activation of MAPK activity
positive regulation of type I hypersensitivity
serotonin secretion
signal transduction
cell surface receptor signaling pathway
activation of JUN kinase activity
leukotriene biosynthetic process
Fc-epsilon receptor signaling pathway
positive regulation of mast cell degranulation
positive regulation of interleukin-3 biosynthetic process
positive regulation of granulocyte macrophage colony-stimulating factor biosynthetic process
positive regulation of peptidyl-tyrosine phosphorylation
positive regulation of calcium-mediated signaling
protein binding
IgE receptor activity
IgE binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2120244 | BY585428.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2120244 | J05018.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2120244 | NM_010184.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 536052 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 903654 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 166914 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P173953 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2120244 | BY585428.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2120244 | J05018.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2120244 | NM_010184.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P429770 | BC125457.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P429770 | BC145138.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P429770 | BC125455.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P429770 | J05018.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836869 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836871 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836836 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836885 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836850 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836847 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836874 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836880 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836848 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836853 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836841 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836831 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836833 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836859 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836855 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836858 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836839 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836843 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836837 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836835 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836882 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836845 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836844 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836886 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P173953 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04358385 | - | - |
| Whole Mouse Genome G4122A | A_51_P429770 | BC125457.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P429770 | BC145138.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P429770 | BC125455.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P429770 | J05018.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102969 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P173953 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00831506 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00831492 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00831496 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00323895 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00831505 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00323897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20836751 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836866 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836829 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836809 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836872 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836792 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836800 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00831493 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836869 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836826 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836807 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836833 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836863 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00831505 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836850 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836861 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00831497 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836874 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836855 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836880 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836886 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836814 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836882 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836805 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836801 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836896 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00307688 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00307687 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00323900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836836 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836794 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836789 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836890 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836845 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836797 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836867 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836831 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836803 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836837 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836818 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836841 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836854 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836853 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00136383 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01116973 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836812 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836879 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836852 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836827 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836835 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00283524 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836791 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836819 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836844 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00670595 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00303817 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836832 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836815 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01116972 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836813 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836897 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836810 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836894 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10360341 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360343 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360342 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360340 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360339 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421775_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10360338 | - | - |
| Mouse Genome 430 2.0 Array | 1421775_at | - | - |
| Mouse Genome 430 2.0 Array | 1421775_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14125_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14125_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100029397.mm.1 | NM_010184.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100029397.mm.1 | NM_010184.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100029397.mm.1 | TC0100003584.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1421775_at | - | - |
| Mouse Expression Set 430 | 1421775_PM_at | - | - |
| Murine Genome U74v2 Set | 101209_PM_at | - | - |
| Murine Genome U74v2 Set | 101209_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421775_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421775_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421775_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421775_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 761054 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1050671 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001050671 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2771342 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2771342 | - | - |
| mouse-6 v1.1 (Illumina) | scl014125.2_20-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1050671 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl014125.2_20-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1050671 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2863390 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2771342 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2863390 | - | - |

Fcer1a
Genomic Map