Assay Details
| Gene Symbol: Fgf14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010201.4 | NP_034331.2 | 1 - 2 | 219 | 82 | |
| GenBank mRNA | AK030616.1 | - | 1 - 2 | 578 | 82 | |
| U66204.1 | - | 1 - 2 | 194 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fgf14
Genomic Map