Assay Details
| Gene Symbol: Fgf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166361.1 | NP_001159833.1 | 4 - 5 | 537 | 66 | |
| NM_001166362.1 | NP_001159834.1 | 3 - 4 | 450 | 66 | ||
| NM_001166363.1 | NP_001159835.1 | 3 - 4 | 417 | 66 | ||
| NM_010205.2 | NP_034335.1 | 3 - 4 | 609 | 66 | ||
| GenBank mRNA | AK131980.1 | - | 3 - 4 | 426 | 66 | |
| BC048734.1 | - | 4 - 5 | 527 | 6513131 | 66 | |
| BU512456.1 | - | 3 - 4 | 485 | 66 | ||
| D12482.1 | - | 3 - 4 | 423 | 66 | ||
| D12483.1 | - | 3 - 4 | 608 | 66 | ||
| HQ260698.1 | - | 2 - 3 | 224 | 66 | ||
| U18673.1 | - | 3 - 4 | 301 | 66 | ||
| Z48746.1 | - | 3 - 4 | 277 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fgf8
Genomic Map