Assay Details
| Gene Symbol: Fgfr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079908.2 | NP_001073377.1 | 2 - 3 | 76 | ||
| NM_010206.3 | NP_034336.2 | 2 - 3 | 76 | |||
| GenBank mRNA | AK049704.1 | - | 2 - 3 | 76 | ||
| BC010200.1 | - | 2 - 3 | 750 | 3967874 | 76 | |
| M28998.1 | - | 1 - 2 | 149 | 76 | ||
| U22324.1 | - | 1 - 2 | 149 | 76 | ||
| X57790.1 | - | 1 - 2 | 92 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fgfr1
Genomic Map