Assay Details
| Gene Symbol: Figf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC080770.1 | - | 3 - 4 | 776 | 30286444 | 69 |
| D89628.1 | - | 3 - 4 | 608 | 69 | ||
| X99572.1 | - | 3 - 4 | 795 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Figf
Genomic Map