Assay Details
| Gene Symbol: Fmn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285458.1 | NP_001272387.1 | 12 - 13 | 3795 | 91 | |
| NM_001285459.1 | NP_001272388.1 | 12 - 13 | 3419 | 91 | ||
| NM_010230.2 | NP_034360.2 | 15 - 16 | 4197 | 91 | ||
| GenBank mRNA | BC138036.1 | - | 11 - 12 | 3701 | 8861055 | 91 |
| BC171945.1 | - | 11 - 12 | 3695 | 9054231 | 91 | |
| CO814739.2 | - | 7 - 8 | 646 | 91 | ||
| X53599.1 | - | 15 - 16 | 4157 | 91 | ||
| X62379.1 | - | 12 - 13 | 3425 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fmn1
Genomic Map