Assay Details
| Gene Symbol: Gal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010253.3 | NP_034383.1 | 2 - 3 | 223 | 81 | |
| GenBank mRNA | AK133522.1 | - | 2 - 3 | 239 | 81 | |
| BC044055.1 | - | 2 - 3 | 223 | 6476459 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gal
Genomic Map