Assay Details
| Gene Symbol: Gck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287386.1 | NP_001274315.1 | 4 - 5 | 608 | 72 | |
| NM_010292.5 | NP_034422.2 | 4 - 5 | 966 | 72 | ||
| GenBank mRNA | AK142698.1 | - | 4 - 5 | 607 | 72 | |
| BC011139.1 | - | 4 - 5 | 597 | 4195943 | 72 | |
| L38990.1 | - | 4 - 5 | 484 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gck
Genomic Map