Assay Details
| Gene Symbol: Hc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010406.2 | NP_034536.1 | 35 - 36 | 4463 | 70 | |
| GenBank mRNA | BC003963.1 | - | 6 - 7 | 507 | 70 | |
| M35525.1 | - | 35 - 36 | 4418 | 70 | ||
| M35526.1 | - | 35 - 36 | 4416 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hc
Genomic Map