Assay Details
| Gene Symbol: Hc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010406.2 | NP_034536.1 | 39 - 40 | 4834 | 62 | |
| GenBank mRNA | BC003963.1 | - | 10 - 11 | 878 | 62 | |
| M35525.1 | - | 39 - 40 | 4789 | 62 | ||
| M35526.1 | - | 39 - 40 | 4787 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hc
Genomic Map