Assay Details
| Gene Symbol: Hoxb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010461.2 | NP_034591.1 | 1 - 2 | 1485 | 82 | |
| GenBank mRNA | BC139209.1 | - | 1 - 2 | 665 | 8862231 | 82 |
| BC139212.1 | - | 2 - 3 | 605 | 8862234 | 82 | |
| X13721.1 | - | 1 - 2 | 1484 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
grooming behavior
adult locomotory behavior
anterior/posterior pattern specification
sensory perception of pain
dorsal spinal cord development
negative regulation of myeloid cell differentiation
embryonic skeletal system morphogenesis
skeletal system morphogenesis
DNA binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
grooming behavior
adult locomotory behavior
anterior/posterior pattern specification
sensory perception of pain
dorsal spinal cord development
negative regulation of myeloid cell differentiation
embryonic skeletal system morphogenesis
skeletal system morphogenesis
DNA binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28043435 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28043440 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28043433 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28043441 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28043442 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28043432 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28043444 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28043443 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28043434 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2024948 | NM_010461.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2024948 | X06762.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 52087 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 176434 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 618822 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 100069 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2024948 | NM_010461.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2024948 | X06762.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P402686 | X13721.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P312043 | X13721.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P312043 | NM_010461.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P312043 | AK083980.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02402375 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02402369 | - | - |
| Whole Mouse Genome G4122A | A_52_P312043 | X13721.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P312043 | NM_010461.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P312043 | AK083980.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P402686 | X13721.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P02402369 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09700293 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09700310 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09700299 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09007298 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09700300 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09700297 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09700307 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28043444 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28043441 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28043435 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28043443 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28043434 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28043440 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28043442 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28043433 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28043432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28043468 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28043435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28043404 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28043437 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28043425 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09250450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09132186 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28043412 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28043399 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28043429 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28043390 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28043428 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28043422 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09700299 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28043418 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28043409 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28043398 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28043391 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28043388 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09250445 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28043394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09700294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28043416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28043439 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28043410 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09700297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09700300 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28043448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28043413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28043396 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28043407 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28043384 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28043386 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09673952 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09250451 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28043381 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09007298 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28043449 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28043403 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28043430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28043401 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28043433 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28043400 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28043438 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10380632 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380630 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380633 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380631 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10380629 | - | - |
| Mouse Genome 430 2.0 Array | 1452493_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1452493_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15416_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15416_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100013826.mm.1 | NM_010461.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100013826.mm.1 | NM_010461.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100013826.mm.1 | TC1100001493.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100013830.mm.1 | NM_010461.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100013830.mm.1 | NM_010461.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100013830.mm.1 | TC1100001493.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100013829.mm.1 | NM_010461.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100013829.mm.1 | NM_010461.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100013829.mm.1 | TC1100001493.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100013819.mm.1 | NM_010461.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100013819.mm.1 | NM_010461.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100013819.mm.1 | TC1100001493.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100013828.mm.1 | NM_010461.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100013828.mm.1 | NM_010461.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100013828.mm.1 | TC1100001493.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100013816.mm.1 | NM_010461.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100013816.mm.1 | NM_010461.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100013816.mm.1 | TC1100001493.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100013820.mm.1 | NM_010461.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100013820.mm.1 | NM_010461.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100013820.mm.1 | TC1100001493.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100013817.mm.1 | NM_010461.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100013817.mm.1 | NM_010461.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100013817.mm.1 | TC1100001493.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100013818.mm.1 | NM_010461.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100013818.mm.1 | NM_010461.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100013818.mm.1 | TC1100001493.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452493_PM_s_at | - | - |
| Mouse Expression Set 430 | 1452493_s_at | - | - |
| Murine Genome U74v2 Set | 92915_PM_s_at | - | - |
| Murine Genome U74v2 Set | 92915_s_at | - | - |
| Murine Genome U74v2 Set | 96418_PM_r_at | - | - |
| Murine Genome U74v2 Set | 96417_PM_s_at | - | - |
| Murine Genome U74v2 Set | 96417_s_at | - | - |
| Murine Genome U74v2 Set | 96418_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452493_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452493_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 480279 | - | - |
| ABI Mouse Genome Survey Microarray | 726960 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3710450 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3710450 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003710450 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2687621 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2687621 | - | - |
| mouse-6 v1.1 (Illumina) | scl015416.4_119-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3710450 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl015416.4_119-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3710450 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2687621 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2687621 | - | - |

Hoxb8
Genomic Map