Assay Details
| Gene Symbol: Mmp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK148184.1 | - | 8 - 9 | 1632 | 89 | |
| AY622966.1 | - | 1 - 2 | 62 | 89 | ||
| M84324.1 | - | 8 - 9 | 1379 | 89 | ||
More Information
Set Membership: |
|

Mmp2
Genomic Map