Assay Details
| Gene Symbol: Mmp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008610.2 | NP_032636.1 | 9 - 10 | 1777 | 85 | |
| GenBank mRNA | AK148184.1 | - | 9 - 10 | 1776 | 85 | |
| AY622966.1 | - | 2 - 3 | 206 | 85 | ||
| BC070430.1 | - | 9 - 10 | 1777 | 6813184 | 85 | |
| M84324.1 | - | 9 - 10 | 1523 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mmp2
Genomic Map