Assay Details
| Gene Symbol: Mmp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK148184.1 | - | 12 - 13 | 2183 | 103 | |
| M84324.1 | - | 12 - 13 | 1930 | 103 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Mmp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK148184.1 | - | 12 - 13 | 2183 | 103 | |
| M84324.1 | - | 12 - 13 | 1930 | 103 | ||
Set Membership: |
|