Assay Details
| Gene Symbol: Igf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111274.1 | NP_001104744.1 | 3 - 4 | 616 | 69 | |
| NM_010512.4 | NP_034642.2 | 3 - 4 | 698 | 69 | ||
| NM_184052.3 | NP_908941.1 | 3 - 4 | 616 | 69 | ||
| GenBank mRNA | AK038119.1 | - | 3 - 4 | 1699 | 69 | |
| AY878193.1 | - | 3 - 4 | 496 | 69 | ||
| BC012409.1 | - | 2 - 3 | 353 | 4194295 | 69 | |
| CT010364.1 | - | 2 - 3 | 326 | 69 | ||
| X04482.1 | - | 3 - 4 | 398 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Citations (1)
Blood glucose-lowering nuclear receptor agonists only partially normalize hepatic gene expression in db/db mice.
- Authors
- Loffler M, Bilban M, Reimers M, Waldhausl W, Stulnig TM
- Journal
- J Pharmacol Exp Ther - Page No 797 (Wed Feb 01 00:00:00 EST 2006)
- Volume (Year)
- 316 (2006)
- Product Usage
- -

Igf1
Genomic Map