Assay Details
| Gene Symbol: Il4ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008700.3 | NP_001008700.1 | 3 - 4 | 310 | 83 | |
| GenBank mRNA | AF000304.1 | - | 1 - 2 | 87 | 83 | |
| AK034466.1 | - | 3 - 4 | 354 | 83 | ||
| AK042342.1 | - | 3 - 4 | 310 | 83 | ||
| AK088086.1 | - | 3 - 4 | 322 | 83 | ||
| AK137827.1 | - | 3 - 4 | 322 | 83 | ||
| AK149603.1 | - | 3 - 4 | 323 | 83 | ||
| AK149717.1 | - | 3 - 4 | 323 | 83 | ||
| AK150584.1 | - | 3 - 4 | 322 | 83 | ||
| AK151998.1 | - | 3 - 4 | 322 | 83 | ||
| AK153186.1 | - | 3 - 4 | 322 | 83 | ||
| AK159375.1 | - | 3 - 4 | 321 | 83 | ||
| AK160016.1 | - | 3 - 4 | 314 | 83 | ||
| BC112911.1 | - | 3 - 4 | 310 | 30138743 | 83 | |
| BC128301.1 | - | 2 - 3 | 181 | 83 | ||
| BC128302.1 | - | 2 - 3 | 181 | 83 | ||
| M27959.1 | - | 3 - 4 | 305 | 83 | ||
| M27960.1 | - | 3 - 4 | 305 | 83 | ||
| M29854.1 | - | 3 - 4 | 289 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il4ra
Genomic Map