Assay Details
| Gene Symbol: Il4ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008700.3 | NP_001008700.1 | 5 - 6 | 600 | 63 | |
| GenBank mRNA | AF000304.1 | - | 3 - 4 | 377 | 63 | |
| AK034466.1 | - | 5 - 6 | 644 | 63 | ||
| AK042342.1 | - | 5 - 6 | 600 | 63 | ||
| AK088086.1 | - | 5 - 6 | 612 | 63 | ||
| AK137827.1 | - | 5 - 6 | 612 | 63 | ||
| AK149603.1 | - | 5 - 6 | 613 | 63 | ||
| AK149717.1 | - | 5 - 6 | 613 | 63 | ||
| AK150584.1 | - | 5 - 6 | 612 | 63 | ||
| AK151998.1 | - | 5 - 6 | 612 | 63 | ||
| AK153186.1 | - | 5 - 6 | 612 | 63 | ||
| AK159375.1 | - | 5 - 6 | 611 | 63 | ||
| AK160016.1 | - | 5 - 6 | 604 | 63 | ||
| BC112911.1 | - | 5 - 6 | 600 | 30138743 | 63 | |
| BC128301.1 | - | 4 - 5 | 471 | 63 | ||
| BC128302.1 | - | 4 - 5 | 471 | 63 | ||
| M27959.1 | - | 5 - 6 | 595 | 63 | ||
| M27960.1 | - | 5 - 6 | 595 | 63 | ||
| M29854.1 | - | 5 - 6 | 579 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il4ra
Genomic Map