Assay Details
| Gene Symbol: Il6st | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010560.3 | NP_034690.3 | 2 - 3 | 275 | 97 | |
| GenBank mRNA | AK170020.1 | - | 3 - 4 | 437 | 97 | |
| BC020454.1 | - | 2 - 3 | 229 | 97 | ||
| BC058679.1 | - | 2 - 3 | 236 | 6834623 | 97 | |
| M83336.1 | - | 1 - 2 | 80 | 97 | ||
| X62646.1 | - | 1 - 2 | 80 | 97 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK170020.1,BC058679.1,M83336.1,NM_010560.3,X62646.1 transcripts.
Set Membership: |
|

Il6st
Genomic Map