| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100035821.mm.1 |
NM_010575.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100035821.mm.1 |
NM_010575.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100035821.mm.1 |
TC1100003839.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100035826.mm.1 |
NM_010575.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100035826.mm.1 |
NM_010575.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100035826.mm.1 |
TC1100003839.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100035825.mm.1 |
NM_010575.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100035825.mm.1 |
NM_010575.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100035825.mm.1 |
TC1100003839.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100035827.mm.1 |
NM_010575.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100035827.mm.1 |
NM_010575.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100035827.mm.1 |
TC1100003839.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100035823.mm.1 |
NM_010575.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100035823.mm.1 |
NM_010575.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100035823.mm.1 |
TC1100003839.mm.1 |
MTA TC |