Assay Details
| Gene Symbol: Itga2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010575.2 | NP_034705.2 | 2 - 3 | 480 | 66 | |
| GenBank mRNA | AF166384.1 | - | 2 - 3 | 358 | 66 | |
| AF170316.1 | - | 2 - 3 | 341 | 66 | ||
| AK142289.1 | - | 2 - 3 | 342 | 66 | ||
| AK154619.1 | - | 2 - 3 | 481 | 66 | ||
| AK156872.1 | - | 2 - 3 | 343 | 66 | ||
| AK158443.1 | - | 2 - 3 | 327 | 66 | ||
| BC120493.1 | - | 2 - 3 | 330 | 40129416 | 66 | |
| BC137570.1 | - | 2 - 3 | 330 | 8860589 | 66 | |
More Information
Set Membership: |
|

Itga2b
Genomic Map