Assay Details
| Gene Symbol: Itga4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010576.3 | NP_034706.3 | 1 - 2 | 430 | 108 | |
| GenBank mRNA | AB850880.1 | - | 1 - 2 | 195 | 108 | |
| AF109136.1 | - | 1 - 2 | 266 | 108 | ||
| AK039959.1 | - | 1 - 2 | 284 | 108 | ||
| AK051676.1 | - | 1 - 2 | 356 | 108 | ||
| AK078842.1 | - | 1 - 2 | 135 | 108 | ||
| AK156015.1 | - | 1 - 2 | 326 | 108 | ||
| AK156517.1 | - | 1 - 2 | 200 | 108 | ||
| BC068313.1 | - | 1 - 2 | 210 | 30093248 | 108 | |
| X53176.1 | - | 1 - 2 | 353 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Itga4
Genomic Map