Assay Details
| Gene Symbol: Itgb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145884.1 | NP_001139356.1 | 11 - 12 | 2216 | 64 | |
| NM_010580.2 | NP_034710.2 | 11 - 12 | 2216 | 64 | ||
| GenBank mRNA | AF022110.1 | - | 11 - 12 | 2215 | 64 | |
| AF043256.1 | - | 11 - 12 | 2121 | 64 | ||
| AF043257.1 | - | 11 - 12 | 2121 | 64 | ||
| AK087476.1 | - | 11 - 12 | 1963 | 64 | ||
| AK140251.1 | - | 11 - 12 | 2128 | 64 | ||
| AK143532.1 | - | 11 - 12 | 2151 | 64 | ||
| BC029170.1 | - | 6 - 7 | 1063 | 64 | ||
| BC046280.1 | - | 11 - 12 | 2125 | 6394641 | 64 | |
| BC058246.1 | - | 11 - 12 | 2180 | 6313668 | 64 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Itgb5
Genomic Map