Assay Details
| Gene Symbol: Itk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281965.1 | NP_001268894.1 | 15 - 16 | 1753 | 78 | |
| NM_001281966.1 | NP_001268895.1 | 15 - 16 | 1744 | 78 | ||
| NM_001281968.1 | NP_001268897.1 | 15 - 16 | 1732 | 78 | ||
| NM_010583.3 | NP_034713.2 | 15 - 16 | 1735 | 78 | ||
| GenBank mRNA | AK037554.1 | - | 815 | 78 | ||
| AK083380.1 | - | 15 - 16 | 1707 | 78 | ||
| AK153613.1 | - | 15 - 16 | 1691 | 78 | ||
| BC128374.1 | - | 15 - 16 | 1661 | 40111687 | 78 | |
| BC128375.1 | - | 15 - 16 | 1658 | 40111688 | 78 | |
| D14042.1 | - | 15 - 16 | 1675 | 78 | ||
| L00619.1 | - | 15 - 16 | 1740 | 78 | ||
| L05631.1 | - | 15 - 16 | 1701 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Itk
Genomic Map