Assay Details
| Gene Symbol: Itpr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010585.5 | NP_034715.3 | 36 - 37 | 4978 | 58 | |
| GenBank mRNA | CF744794.1 | - | 2 - 3 | 243 | 58 | |
| X15373.1 | - | 36 - 37 | 4956 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Itpr1
Genomic Map