Assay Details
| Gene Symbol: Kcnab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289450.1 | NP_001276379.1 | 8 - 9 | 506 | 97 | |
| NM_010597.4 | NP_034727.3 | 7 - 8 | 922 | 97 | ||
| GenBank mRNA | AF033003.1 | - | 7 - 8 | 97 | ||
| AK004666.1 | - | 8 - 9 | 490 | 97 | ||
| AK015412.1 | - | 9 - 10 | 762 | 97 | ||
| AK031362.1 | - | 7 - 8 | 923 | 97 | ||
| AK138467.1 | - | 7 - 8 | 911 | 97 | ||
| AK153200.1 | - | 8 - 9 | 508 | 97 | ||
| BC014701.1 | - | 7 - 8 | 647 | 4482268 | 97 | |
| U65591.1 | - | 7 - 8 | 97 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Kcnab1
Genomic Map