Assay Details
| Gene Symbol: Kcnab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252654.1 | NP_001239583.1 | 5 - 6 | 444 | 103 | |
| NM_001252655.1 | NP_001239584.1 | 5 - 6 | 473 | 103 | ||
| NM_001252656.1 | NP_001239585.1 | 6 - 7 | 515 | 103 | ||
| NM_010598.3 | NP_034728.2 | 6 - 7 | 486 | 103 | ||
| GenBank mRNA | AK143166.1 | - | 5 - 6 | 421 | 103 | |
| AK152613.1 | - | 6 - 7 | 438 | 103 | ||
| AK157053.1 | - | 5 - 6 | 437 | 103 | ||
| BC039178.1 | - | 6 - 7 | 416 | 4503373 | 103 | |
| L48983.1 | - | 6 - 7 | 478 | 103 | ||
| U31908.1 | - | 6 - 7 | 423 | 103 | ||
| U65592.1 | - | 5 - 6 | 281 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kcnab2
Genomic Map