Assay Details
| Gene Symbol: Kcnab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252654.1 | NP_001239583.1 | 7 - 8 | 536 | 80 | |
| NM_001252655.1 | NP_001239584.1 | 7 - 8 | 565 | 80 | ||
| NM_001252656.1 | NP_001239585.1 | 8 - 9 | 607 | 80 | ||
| NM_010598.3 | NP_034728.2 | 8 - 9 | 578 | 80 | ||
| GenBank mRNA | AK143166.1 | - | 7 - 8 | 513 | 80 | |
| AK152613.1 | - | 8 - 9 | 530 | 80 | ||
| AK157053.1 | - | 7 - 8 | 529 | 80 | ||
| BC039178.1 | - | 8 - 9 | 508 | 4503373 | 80 | |
| L48983.1 | - | 8 - 9 | 570 | 80 | ||
| U31908.1 | - | 8 - 9 | 515 | 80 | ||
| U65592.1 | - | 7 - 8 | 373 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kcnab2
Genomic Map