Assay Details
| Gene Symbol: Ldlr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252658.1 | NP_001239587.1 | 7 - 8 | 1264 | 97 | |
| NM_001252659.1 | NP_001239588.1 | 7 - 8 | 1261 | 97 | ||
| NM_010700.3 | NP_034830.2 | 7 - 8 | 1264 | 97 | ||
| GenBank mRNA | AF425607.1 | - | 7 - 8 | 1112 | 97 | |
| AK076243.1 | - | 7 - 8 | 1242 | 97 | ||
| AK149950.1 | - | 7 - 8 | 1202 | 97 | ||
| AK151968.1 | - | 7 - 8 | 1231 | 97 | ||
| AK152103.1 | - | 7 - 8 | 1200 | 97 | ||
| AK160006.1 | - | 7 - 8 | 1202 | 97 | ||
| AK161104.1 | - | 7 - 8 | 1202 | 97 | ||
| BC053041.1 | - | 7 - 8 | 1209 | 5704871 | 97 | |
| X64414.1 | - | 7 - 8 | 1183 | 97 | ||
| Z19521.1 | - | 7 - 8 | 1062 | 97 | ||
More Information
Set Membership: |
|

Ldlr
Genomic Map