Assay Details
| Gene Symbol: Lepr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122899.1 | NP_001116371.1 | 13 - 14 | 2541 | 119 | |
| NM_010704.2 | NP_034834.1 | 13 - 14 | 2541 | 119 | ||
| NM_146146.2 | NP_666258.2 | 13 - 14 | 2541 | 119 | ||
| GenBank mRNA | AK140785.1 | - | 13 - 14 | 2043 | 119 | |
| AK143989.1 | - | 13 - 14 | 2230 | 119 | ||
| U42467.1 | - | 13 - 14 | 2049 | 119 | ||
| U46135.1 | - | 13 - 14 | 2049 | 119 | ||
| U49106.1 | - | 12 - 13 | 1989 | 119 | ||
| U49109.1 | - | 12 - 13 | 1989 | 119 | ||
| U49110.1 | - | 12 - 13 | 1989 | 119 | ||
| U52915.1 | - | 13 - 14 | 2503 | 119 | ||
| U58861.1 | - | 13 - 14 | 2071 | 119 | ||
| U58862.1 | - | 13 - 14 | 2081 | 119 | ||
| U58863.1 | - | 13 - 14 | 2071 | 119 | ||
| Y10296.1 | - | 12 - 13 | 1997 | 119 | ||
| Y10298.1 | - | 12 - 13 | 1997 | 119 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lepr
Genomic Map