Assay Details
| Gene Symbol: Limk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK141813.1 | - | 2 - 3 | 333 | 68 | |
| AK164484.1 | - | 2 - 3 | 334 | 68 | ||
| BC063768.1 | - | 2 - 3 | 315 | 68 | ||
| CV563459.1 | - | 1 - 2 | 92 | 68 | ||
| U14166.1 | - | 2 - 3 | 356 | 68 | ||
| U15159.1 | - | 2 - 3 | 68 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Limk1
Genomic Map