Assay Details
| Gene Symbol: Limk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK141813.1 | - | 3 - 4 | 470 | 66 | |
| AK164484.1 | - | 3 - 4 | 471 | 66 | ||
| AK170166.1 | - | 2 - 3 | 245 | 66 | ||
| AK172083.1 | - | 2 - 3 | 229 | 66 | ||
| BC063768.1 | - | 3 - 4 | 452 | 66 | ||
| BY230595.1 | - | 2 - 3 | 272 | 66 | ||
| CV563459.1 | - | 2 - 3 | 229 | 66 | ||
| U14166.1 | - | 3 - 4 | 493 | 66 | ||
| U15159.1 | - | 3 - 4 | 468 | 66 | ||
More Information
Set Membership: |
|

Limk1
Genomic Map