Assay Details
| Gene Symbol: Limk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK139231.1 | - | 1 - 2 | 2590 | 62 | |
| AK141813.1 | - | 8 - 9 | 1245 | 62 | ||
| AK170166.1 | - | 7 - 8 | 1020 | 62 | ||
| AK172083.1 | - | 7 - 8 | 1004 | 62 | ||
| BC063768.1 | - | 8 - 9 | 1225 | 62 | ||
| U14166.1 | - | 8 - 9 | 1268 | 62 | ||
| U15159.1 | - | 8 - 9 | 1240 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK139231.1 transcripts.
Set Membership: |
|

Limk1
Genomic Map