Assay Details
| Gene Symbol: Psmb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010724.2 | NP_034854.2 | 2 - 3 | 500 | 67 | |
| GenBank mRNA | AK031801.1 | - | 2 - 3 | 325 | 67 | |
| AK050627.1 | - | 2 - 3 | 328 | 67 | ||
| AK146182.1 | - | 2 - 3 | 347 | 67 | ||
| AK150674.1 | - | 2 - 3 | 328 | 67 | ||
| AK159971.1 | - | 2 - 3 | 317 | 67 | ||
| AK169753.1 | - | 1 - 2 | 773 | 67 | ||
| BC013785.1 | - | 2 - 3 | 324 | 2654130 | 67 | |
| BC051450.1 | - | 2 - 3 | 331 | 30076739 | 67 | |
| KJ650228.1 | - | 2 - 3 | 296 | 67 | ||
| U22031.1 | - | 2 - 3 | 296 | 67 | ||
| U22032.1 | - | 2 - 3 | 296 | 67 | ||
| U22033.1 | - | 2 - 3 | 296 | 67 | ||
| U22034.1 | - | 2 - 3 | 296 | 67 | ||
| U22035.1 | - | 2 - 3 | 296 | 67 | ||
| X64449.1 | - | 2 - 3 | 315 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Psmb8
Genomic Map