Assay Details
| Gene Symbol: Ltbr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010736.3 | NP_034866.1 | 4 - 5 | 657 | 81 | |
| GenBank mRNA | AK146127.1 | - | 4 - 5 | 640 | 81 | |
| BC138590.1 | - | 4 - 5 | 520 | 8861610 | 81 | |
| BC138591.1 | - | 4 - 5 | 520 | 8861611 | 81 | |
| L38423.1 | - | 4 - 5 | 657 | 81 | ||
| U29173.1 | - | 4 - 5 | 641 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ltbr
Genomic Map